missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane, 98+%, stabilized, Thermo Scientific Chemicals

£39.20 - £146.73

Chemical Identifiers

CAS 75-30-9, 7440-50-8
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001071
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
PubChem CID 6362
IUPAC Name 2-iodopropane
SMILES CC(C)I
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10011880
View Documents Promotion Details
Thermo Scientific Acros
164961000
100 mL Glass bottle
£39.20
100mL
Please to purchase this item. Need a web account? Register with us today!
10124180
View Documents
Thermo Scientific
164965000
500 mL Glass bottle
£146.73
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-30-9, 7440-50-8
169.99
FMKOJHQHASLBPH-UHFFFAOYSA-N
6362
CC(C)I
C3H7I
MFCD00001071
isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
2-iodopropane

Specifications

75-30-9, 7440-50-8
100.0
Colorless to Yellow
90.0°C
Authentic
C3H7I
1.4972 to 1.4992
01, 114
1.7
Solubility in water: slightly soluble. Other solubilities: miscible with chloroform,ehter,alcohol and,benzene
CC(C)I
copper
6362
98+%
2-Iodopropane, 98+%
98.0
-90.0°C
1.7000g/mL
42°C
98% min. (GC)
(CH3)2CHI
MFCD00001071
15, 5260
isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
FMKOJHQHASLBPH-UHFFFAOYSA-N
2-iodopropane
169.99
169.99
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 200-859-3

missing translation for 'rtecsNumber' : TZ4200000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only