missing translation for 'onlineSavingsMsg'
Learn More

2-Iodosobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11945141
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304-91-6
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.02
InChI Key IFPHDUVGLXEIOQ-UHFFFAOYSA-N
Synonym 2-iodosobenzoic acid, o-iodosobenzoic acid, o-iodosylbenzoic acid, o-iodosobenzoate, benzoic acid, 2-iodosyl, benzoic acid, o-iodoso, 2-iodosyl benzoate, benzoic acid, iodosyl, ortho-iodosylbenzoic acid, 3h-1,2-benziodoxol-1-ium, 3-oxo-, hydroxide
PubChem CID 67537
ChEBI CHEBI:52698
IUPAC Name 2-iodosylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I=O

Specifications

Melting Point 200.0°C to 204.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Iodometry)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 264.02
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Iodosobenzoic acid
compliance-icons
Product Identifier
  • 2-Iodosobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'