missing translation for 'onlineSavingsMsg'
Learn More
2-Isobutyl-3-methoxypyrazine, 98%, Thermo Scientific Chemicals Product Code.: 11494644

2-Isobutyl-3-methoxypyrazine, 98%, Thermo Scientific Chemicals

Product Code. 11494644
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11494644

Brand: Thermo Scientific Alfa Aesar A13680.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24683-00-9
Molecular Formula C9H14N2O
Molecular Weight (g/mol) 166.22
MDL Number MFCD00006128
InChI Key UXFSPRAGHGMRSQ-UHFFFAOYSA-N
Synonym 2-isobutyl-3-methoxypyrazine, 3-isobutyl-2-methoxypyrazine, 2-methoxy-3-2-methylpropyl pyrazine, 2-methoxy-3-isobutylpyrazine, 2-methoxy-3-isobutyl pyrazine, 1dzk, pyrazine, 2-methoxy-3-2-methylpropyl, unii-s327o0t12o, ibmp, 3-methoxy-2-isobutylpyrazineShow MoreShow Less
PubChem CID 32594
IUPAC Name 2-methoxy-3-(2-methylpropyl)pyrazine
SMILES COC1=NC=CN=C1CC(C)C

Specifications

Density 0.986
Flash Point 80°C (176°F)
Refractive Index 1.49
Quantity 5 g
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 2-Isobutyl-3-methoxypyrazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251503 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13680.06,A13680.03
Lot Number10248632 Certificate TypeCertificate of Analysis Date24/11/2023 Catalog NumberA13680.06,A13680.03,A13680.14
Lot Number10210962 Certificate TypeCertificate of Analysis Date24/05/2023 Catalog NumberA13680.06,A13680.03
Lot NumberH3010A Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13680.06,A13680.03
Lot Number10210962 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13680.06,A13680.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Isobutyl-3-methoxypyrazine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.