missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isocyanatobenzonitrile, 97%, Thermo Scientific™
£47.20 - £416.00
Chemical Identifiers
CAS | 42066-86-4 |
---|---|
Molecular Formula | C8H4N2O |
Molecular Weight (g/mol) | 144.133 |
InChI Key | FQLNVWAFXQUXGW-UHFFFAOYSA-N |
PubChem CID | 2735873 |
IUPAC Name | 2-isocyanatobenzonitrile |
SMILES | C1=CC=C(C(=C1)C#N)N=C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10548253
|
Thermo Scientific Maybridge
BTB13667CB |
250 mg |
£47.20
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10502632
|
Thermo Scientific Maybridge
BTB13667DA |
1 g |
£151.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10066664
|
Thermo Scientific Maybridge
BTB13667DE |
5 g |
£416.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
42066-86-4 | |
144.133 | |
2735873 | |
C1=CC=C(C(=C1)C#N)N=C=O |
C8H4N2O | |
FQLNVWAFXQUXGW-UHFFFAOYSA-N | |
2-isocyanatobenzonitrile |
Specifications
42066-86-4 | |
Amber glass bottle | |
CNC6H4NCO | |
FQLNVWAFXQUXGW-UHFFFAOYSA-N | |
2-isocyanatobenzonitrile | |
2735873 | |
95% |
95% | |
C8H4N2O | |
2-cyanophenyl isocyanate, benzonitrile,2-isocyanato, 2-isocyanato-benzonitrile, 2-isocyanatobenzenecarbonitrile, 2-cyanophenylisocyanate, benzonitrile, 2-isocyanato, acmc-1ap1b | |
C1=CC=C(C(=C1)C#N)N=C=O | |
144.133 | |
144.13 | |
2-Isocyanatobenzonitrile |