missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropylphenyl isothiocyanate, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Specifications
CAS Min % | 95.0 |
Density | 1.0500g/mL |
Assay Percent Range | 95%+ |
Packaging | Amber Glass Bottle |
Linear Formula | CH3CH(CH3)C6H4NCS |
Quantity | 1 g |
Formula Weight | 177.27 |
Percent Purity | 95% |
Chemical Name or Material | 2-Isopropylphenyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'