Learn More
2-Ketoglutaric acid, 98%, Thermo Scientific Chemicals
£23.20 - £173.00
Chemical Identifiers
CAS | 328-50-7 |
---|---|
Molecular Formula | C5H6O5 |
Molecular Weight (g/mol) | 146.098 |
MDL Number | MFCD00004165 |
InChI Key | KPGXRSRHYNQIFN-UHFFFAOYSA-N |
Synonym | 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid |
PubChem CID | 51 |
ChEBI | CHEBI:30915 |
IUPAC Name | 2-oxopentanedioic acid |
SMILES | C(CC(=O)O)C(=O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493963
|
Thermo Scientific Alfa Aesar
A10256.14 |
25 g |
£23.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403973
|
Thermo Scientific Alfa Aesar
A10256.22 |
100 g |
£54.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413973
|
Thermo Scientific Alfa Aesar
A10256.36 |
500 g |
£173.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Ketoglutaric acid is used as an intermediate in the kreb’s cycle catalyzed by glutamate dehydrogenase enzyme on glutamate. It is also used in dietary supplements to improve protein synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ketoglutaric acid is used as an intermediate in the kreb’s cycle catalyzed by glutamate dehydrogenase enzyme on glutamate. It is also used in dietary supplements to improve protein synthesis.
Solubility
Soluble in water, methanol and dimethyl sulfoxide.
Notes
Store away from heat store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
328-50-7 | |
146.098 | |
KPGXRSRHYNQIFN-UHFFFAOYSA-N | |
51 | |
2-oxopentanedioic acid |
C5H6O5 | |
MFCD00004165 | |
2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid | |
CHEBI:30915 | |
C(CC(=O)O)C(=O)C(=O)O |
Specifications
328-50-7 | |
C5H6O5 | |
1705689 | |
2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid | |
KPGXRSRHYNQIFN-UHFFFAOYSA-N | |
2-oxopentanedioic acid | |
51 | |
146.1 | |
2-Ketoglutaric acid |
113°C to 117°C | |
MFCD00004165 | |
14,5303 | |
Soluble in water,methanol and dimethyl sulfoxide. | |
C(CC(=O)O)C(=O)C(=O)O | |
146.098 | |
CHEBI:30915 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P280-P305+P351+P338-P310
H318
missing translation for 'einecsNumber' : 206-330-3
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only