missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzothiazole, ≥99.0%, Honeywell Fluka™

Product Code. 15606100
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

for the determination of Ag, Au, Bi, Cd, Hg, Ir, Pt, Tl,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-30-4
Molecular Formula C7H5NS2
Molecular Weight (g/mol) 167.244
MDL Number MFCD00005781
InChI Key YXIWHUQXZSMYRE-UHFFFAOYSA-N
Synonym 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
PubChem CID 697993
ChEBI CHEBI:34292
IUPAC Name 3H-1,3-benzothiazole-2-thione
SMILES C1=CC=C2C(=C1)NC(=S)S2

Specifications

Melting Point 179°C to 182°C
Color Yellow
Boiling Point 260°C
Odor No data
Packaging Poly bottle
Quantity 250 g
UN Number UN3077
Beilstein 119484
Formula Weight 167.25g/mol
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material 2-Mercaptobenzothiazole
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'