missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptobenzothiazole, ≥99.0%, Honeywell Fluka™
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 179°C to 182°C |
Color | Yellow |
Boiling Point | 260°C |
Odor | No data |
Packaging | Poly bottle |
Quantity | 50 g |
UN Number | UN3077 |
Beilstein | 119484 |
Formula Weight | 167.25g/mol |
Percent Purity | ≥99.0% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'