missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzyl alcohol, tech. 90%, Thermo Scientific Chemicals

Product Code. 11342477
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4521-31-7
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00014448
InChI Key FYWFCRHZXORPFH-UHFFFAOYSA-N
Synonym 2-mercaptobenzyl alcohol, 2-mercaptophenyl methanol, o-mercaptobenzyl alcohol, 2-sulfanylphenyl methanol, 2-mercaptobenzylalcohol, 2-hydroxymethyl thiophenol, benzenemethanol, 2-mercapto, hydroxymethylthiophenol, o-mercaptobenzylalcohol, acmc-1csoz
PubChem CID 138278
IUPAC Name (2-sulfanylphenyl)methanol
SMILES C1=CC=C(C(=C1)CO)S

Specifications

Melting Point 31°C to 32°C
Density 1.21
Boiling Point 94°C to 96°C (0.2 mmHg)
Flash Point >110°C (230°F)
Odor Like Rotten Eggs
Refractive Index 1.623
Quantity 25 g
Beilstein 2206253
Formula Weight 140.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Mercaptobenzyl alcohol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'