missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mesitylethanol, 98%, Thermo Scientific™
£20.50 - £70.20
Chemical Identifiers
CAS | 6950-92-1 |
---|---|
Molecular Formula | C11H16O |
Molecular Weight (g/mol) | 164.25 |
MDL Number | MFCD00020609 |
InChI Key | FQZPTDPHVFTOSY-UHFFFAOYSA-N |
Synonym | 2-mesitylethanol, 2-hydroxyethylmesitylene, 2-hydroxyethyl mesitylene, 2-2,4,6-trimethylphenyl ethanol, 2-2,4,6-trimethylphenyl ethan-1-ol, 2,4,6-trimethylphenethylalcohol, 2-mesitylethan-1-ol, acmc-20ans7, 2,4,6-trimethylbenzeneethanol, 2,4,6-trimethylphenethyl alcohol |
PubChem CID | 81389 |
IUPAC Name | 2-(2,4,6-trimethylphenyl)ethanol |
SMILES | CC1=CC(C)=C(CCO)C(C)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387955
|
Thermo Scientific Alfa Aesar
L08032.03 |
1 g | |||||||
11397955
|
Thermo Scientific Alfa Aesar
L08032.06 |
5 g |
£70.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
6950-92-1 | |
164.25 | |
FQZPTDPHVFTOSY-UHFFFAOYSA-N | |
81389 | |
CC1=CC(C)=C(CCO)C(C)=C1 |
C11H16O | |
MFCD00020609 | |
2-mesitylethanol, 2-hydroxyethylmesitylene, 2-hydroxyethyl mesitylene, 2-2,4,6-trimethylphenyl ethanol, 2-2,4,6-trimethylphenyl ethan-1-ol, 2,4,6-trimethylphenethylalcohol, 2-mesitylethan-1-ol, acmc-20ans7, 2,4,6-trimethylbenzeneethanol, 2,4,6-trimethylphenethyl alcohol | |
2-(2,4,6-trimethylphenyl)ethanol |
Specifications
6950-92-1 | |
C11H16O | |
1938400 | |
FQZPTDPHVFTOSY-UHFFFAOYSA-N | |
2-(2,4,6-trimethylphenyl)ethanol | |
81389 | |
98% |
80°C to 84°C | |
MFCD00020609 | |
2-mesitylethanol, 2-hydroxyethylmesitylene, 2-hydroxyethyl mesitylene, 2-2,4,6-trimethylphenyl ethanol, 2-2,4,6-trimethylphenyl ethan-1-ol, 2,4,6-trimethylphenethylalcohol, 2-mesitylethan-1-ol, acmc-20ans7, 2,4,6-trimethylbenzeneethanol, 2,4,6-trimethylphenethyl alcohol | |
CC1=CC(C)=C(CCO)C(C)=C1 | |
164.25 | |
164.25 | |
2-Mesitylethanol |
Safety and Handling
missing translation for 'einecsNumber' : 230-123-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures