missing translation for 'onlineSavingsMsg'
Learn More

2'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals

£31.66 - £111.56

Chemical Identifiers

CAS 579-74-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008725
InChI Key DWPLEOPKBWNPQV-UHFFFAOYSA-N
Synonym 2'-methoxyacetophenone, 1-2-methoxyphenyl ethanone, o-methoxyacetophenone, ethanone, 1-2-methoxyphenyl, o-acetylanisole, 1-2-methoxyphenyl ethan-1-one, o-acetanisole, 2-acetylanisole, 2-methyoxyacetophenone oxime, ortho-methoxyacetophenone
PubChem CID 68481
IUPAC Name 1-(2-methoxyphenyl)ethanone
SMILES COC1=CC=CC=C1C(C)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11433914
View Documents
Thermo Scientific Alfa Aesar
A11929.14
25 g
£31.66
25g
Please to purchase this item. Need a web account? Register with us today!
11443914
View Documents
Thermo Scientific Alfa Aesar
A11929.22
100 g
£111.56
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

579-74-8
150.18
DWPLEOPKBWNPQV-UHFFFAOYSA-N
68481
COC1=CC=CC=C1C(C)=O
C9H10O2
MFCD00008725
2'-methoxyacetophenone, 1-2-methoxyphenyl ethanone, o-methoxyacetophenone, ethanone, 1-2-methoxyphenyl, o-acetylanisole, 1-2-methoxyphenyl ethan-1-one, o-acetanisole, 2-acetylanisole, 2-methyoxyacetophenone oxime, ortho-methoxyacetophenone
1-(2-methoxyphenyl)ethanone

Specifications

579-74-8
119°C to 120°C (10 mmHg)
C9H10O2
MFCD00008725
Light sensitive
DWPLEOPKBWNPQV-UHFFFAOYSA-N
1-(2-methoxyphenyl)ethanone
68481
98%
1.09
108°C (226°F)
1.5393
1100602
2'-methoxyacetophenone, 1-2-methoxyphenyl ethanone, o-methoxyacetophenone, ethanone, 1-2-methoxyphenyl, o-acetylanisole, 1-2-methoxyphenyl ethan-1-one, o-acetanisole, 2-acetylanisole, 2-methyoxyacetophenone oxime, ortho-methoxyacetophenone
COC1=CC=CC=C1C(C)=O
150.18
150.18
2'-Methoxyacetophenone
Videos
Safety and Handling

Safety and Handling

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c

H302-H315-H319

missing translation for 'einecsNumber' : 209-446-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 2'-Methoxyacetophenone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : 2'-Methoxyacetophenone

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only