missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzoyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11399779
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21615-34-9
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00000664
InChI Key RZNHSEZOLFEFGB-UHFFFAOYSA-N
Synonym o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi
PubChem CID 88969
IUPAC Name 2-methoxybenzoyl chloride
SMILES COC1=CC=CC=C1C(=O)Cl

Specifications

Density 1.146
Boiling Point 128°C to 129°C (8 mmHg)
Flash Point 84°C (184°F)
Refractive Index 1.5724
Quantity 5 g
UN Number UN1729
Beilstein 607605
Sensitivity Moisture sensitive
Formula Weight 170.6
Percent Purity 97%
Chemical Name or Material 2-Methoxybenzoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'