missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 10215130
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-16-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004611
InChI Key WYLYBQSHRJMURN-UHFFFAOYSA-N
Synonym 2-methoxybenzyl alcohol, 2-methoxyphenyl methanol, o-methoxybenzyl alcohol, benzenemethanol, 2-methoxy, o-anisyl alcohol, 2-methoxybenzylalcohol, 2-methoxybenzenemethanol, unii-457nq8ngcg, 457nq8ngcg, 2-methoxyphenyl methan-1-ol
PubChem CID 69154
IUPAC Name (2-methoxyphenyl)methanol
SMILES COC1=CC=CC=C1CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown-Orange to Colorless
Density 1.0300g/mL
Boiling Point 248°C to 250°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CH2OH
Refractive Index 1.5460 to 1.548
Quantity 10 mL
Beilstein 06, 893
Specific Gravity 1.03
Formula Weight 138.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methoxybenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'