Learn More
2-Methoxycarbonylphenylboronic acid, 97%, Thermo Scientific Chemicals
£29.20 - £94.50
Chemical Identifiers
CAS | 374538-03-1 |
---|---|
Molecular Formula | C8H9BO4 |
Molecular Weight (g/mol) | 179.97 |
MDL Number | MFCD02179452 |
InChI Key | ODAXNYMENLFYMY-UHFFFAOYSA-N |
Synonym | 2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid |
PubChem CID | 2773496 |
SMILES | COC(=O)C1=CC=CC=C1B(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10077949
|
Thermo Scientific Acros
433910010 |
1 g | Glass bottle |
£29.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10732962
|
Thermo Scientific Acros
433910050 |
5 g | Glass bottle |
£94.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
374538-03-1 | |
179.97 | |
ODAXNYMENLFYMY-UHFFFAOYSA-N | |
2773496 |
C8H9BO4 | |
MFCD02179452 | |
2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid | |
COC(=O)C1=CC=CC=C1B(O)O |
Specifications
374538-03-1 | |
100.0 | |
White | |
96% min. (HPLC) | |
MFCD02179452 | |
ODAXNYMENLFYMY-UHFFFAOYSA-N | |
[2-(methoxycarbonyl)phenyl]boronic acid | |
2773496 | |
97% | |
2-Methoxycarbonylphenylboronic acid |
96.0 | |
101.1°C to 101.4°C | |
Authentic | |
C8H9BO4 | |
2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid | |
COC(=O)C1=CC=CC=C1B(O)O | |
179.97 | |
179.97 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
- 2-Methoxycarbonylphenylboronic acid, 97%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only