missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl phenyl sulfide, 98%, Thermo Scientific™

Product Code. 11318445
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77298-24-9
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.25
MDL Number MFCD00041329
InChI Key ONVZBVAUSAELNQ-UHFFFAOYSA-N
Synonym 2-methoxyethyl phenyl sulfide, 2-methoxyethyl phenyl sulfane, 2-methoxyethyl sulfanyl benzene, 2-methoxyethyl phenylsulfide, 1-methoxy-2-phenylthioethane, 2-phenylthio ethyl methyl ether, benzene, 2-methoxyethyl thio
PubChem CID 4056174
IUPAC Name 2-methoxyethylsulfanylbenzene
SMILES COCCSC1=CC=CC=C1

Specifications

Boiling Point 125°C to 126°C (18 mmHg)
Linear Formula C6H5S(CH2)2OCH3
Refractive Index 1.557
Quantity 5 g
Beilstein 5500488
Formula Weight 168.25
Percent Purity 98%
Chemical Name or Material 2-Methoxyethyl phenyl sulfide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'