missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypropene, 97%, stabilized, Thermo Scientific Chemicals

Product Code. 10636394
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-11-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00014929
InChI Key YOWQWFMSQCOSBA-UHFFFAOYSA-N
Synonym 2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b
PubChem CID 8300
IUPAC Name 2-methoxyprop-1-ene
SMILES CC(=C)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.7800g/mL
Boiling Point 35°C to 36°C
Flash Point -29°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(OCH3)=CH2
Refractive Index 1.3810 to 1.384
Quantity 2.5 L
Beilstein 01, IV, 2075
Fieser 02,230; 05,360; 11,329; 12,317
Merck Index 15, 6073
Specific Gravity 0.78
Solubility Information Solubility in water: negligible
Stabilizer 0.5% max. potassium carbonate
Formula Weight 72.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methoxypropene, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methoxypropene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash thoroughly after handling.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'