missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine, 98%, Thermo Scientific Chemicals

Product Code. 10743393
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1628-89-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006262
InChI Key IWTFOFMTUOBLHG-UHFFFAOYSA-N
Synonym pyridine, 2-methoxy, 2-methoxy pyridine, pyridine, methoxy, 2-methoxy-pyridine, unii-geb38j8108, methyl 2-pyridyl ether, methoxypyridine, 6-methoxypyridine, methyl pyridinyl ether, pubchem13201
PubChem CID 74201
IUPAC Name 2-methoxypyridine
SMILES COC1=CC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 142°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)OCH3
Refractive Index 1.5010 to 1.503
Quantity 5 g
Beilstein 21, 44
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,acetone and ether
Formula Weight 109.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methoxypyridine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'