missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxythiobenzamide, 97%, Thermo Scientific Chemicals

Product Code. 15405497
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42590-97-6
Molecular Formula C8H9NOS
Molecular Weight (g/mol) 167.226
MDL Number MFCD04973324
InChI Key YNEHBLLUZJTDOL-UHFFFAOYSA-N
PubChem CID 2759731
IUPAC Name 2-methoxybenzenecarbothioamide
SMILES COC1=CC=CC=C1C(=S)N

Specifications

Melting Point 109°C to 113°C
Quantity 1 g
Synonym 2-methoxythiobenzamide, 2-methoxybenzothioamide, 2-methoxy-thiobenzamide, 2-methoxybenzene-1-carbothioamide, amino 2-methoxyphenyl methane-1-thione, o-methoxythiobenzamide, 2-methoxy thiobenzamide, #, benzenecarbothioamide,2-methoxy, benzenecarbothioamide, 2-methoxy
Formula Weight 167.23
Percent Purity 97%
Chemical Name or Material 2-Methoxythiobenzamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'