missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1,3-dithiane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.212 |
Boiling Point | 78°C to 80°C (9 mmHg) |
Flash Point | 82°C (179°F) |
Refractive Index | 1.5615 |
Quantity | 25 g |
Beilstein | 102766 |
Solubility Information | Not miscible or difficult to mix in water. Soluble in alcohol. |
Formula Weight | 134.25 |
Percent Purity | 98% |
Chemical Name or Material | 2-Methyl-1,3-dithiane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'