missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1,3-thiazole-4-carbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Synonym | 4-formyl-2-methylthiazole, 2-methylthiazole-4-carbaldehyde, 2-methylthiazole-4-carboxaldehyde, 4-thiazolecarboxaldehyde, 2-methyl, 2-methyl-4-thiazolecarboxaldehyde, 2-methyl-4-formylthiazole, 2-methyl-4-fomylthiazole, 4-formyl-2-methyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-methyl, 2-methyl-1,3-thiazole-4-carboxaldehyde |
Formula Weight | 127.17 |
Percent Purity | 97% |
Chemical Name or Material | 2-Methyl-1, 3-thiazole-4-carbaldehyde |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction