missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-2-Methyl-1-butanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.
Solubility
Soluble in water. [183 g/L (0 C)] and alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.
Solubility
Soluble in water. [183 g/L (0 C)] and alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Melting Point | <-70°C |
Density | 0.815 |
Boiling Point | 129°C to 131°C |
Flash Point | 43°C (109°F) |
Refractive Index | 1.411 |
Quantity | 100 mL |
UN Number | UN1105 |
Beilstein | 1718810 |
Merck Index | 14,6030 |
Solubility Information | Soluble in water. [183g/L (0 C)] and alcohol. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'