Learn More
2-Methyl-1-butene, 98%
Description
2-Methyl-1-butene is used as a solvent in organic synthesis. It is also used in the preparation of pinacolone, flavor enhancer, spice, crop protectants and tertiary amyl phenol. Further, it serves as a photosensitive material and concrete dispersant. In addition to this, it is used as a fuel and fuel additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Methyl-1-butene is used as a solvent in organic synthesis. It is also used in the preparation of pinacolone, flavor enhancer, spice, crop protectants and tertiary amyl phenol. Further, it serves as a photosensitive material and concrete dispersant. In addition to this, it is used as a fuel and fuel additive.
Solubility
Soluble in ether, ethanol and benzene. Insoluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -137°C |
| Density | 0.651 |
| Boiling Point | 31°C to 32°C |
| Flash Point | −48°C (−54°F) |
| Refractive Index | 1.374 |
| Quantity | 25 g |
| UN Number | UN2459 |
| Solubility Information | Soluble in ether,ethanol and benzene. Insoluble in water. |
| Formula Weight | 70.13 |
| Percent Purity | 98% |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.