missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-morpholino-2-propanamine, 97%, Thermo Scientific™

Product Code. 10103602
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6105-75-5
Molecular Formula C8H18N2O
Molecular Weight (g/mol) 158.25
MDL Number MFCD01714129
InChI Key JCXYZQUYVNLCTG-UHFFFAOYSA-N
Synonym 2-methyl-1-morpholinopropan-2-amine, 2-methyl-1-morpholin-4-yl propan-2-amine, 1,1-dimethyl-2-morpholin-4-yl-ethylamine, 2-methyl-1-morpholino-2-propanamine, 4-2-amino-2-methylpropyl morpholine, morpholine, 4-2-amino-2-methylpropyl, morpholine, n-2-aminoisobutyl, 1,1-dimethyl-2-morpholin-4-ylethylamine, 1,1-dimethyl-2-morpholin-4-ylethyl amine, 4-morpholineethanamine, alpha,alpha-dimethyl
PubChem CID 201497
IUPAC Name 2-methyl-1-morpholin-4-ylpropan-2-amine
SMILES CC(C)(N)CN1CCOCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material 2-Methyl-1-morpholino-2-propanamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'