missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-Propanol, ≥99.0%, ACS Reagent, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -108°C |
Density | 0.803g/mL at 25°C |
Boiling Point | 108°C |
Flash Point | 28°C (82.4°F) |
Ignition Point | 801°F |
Packaging | Glass Bottle |
Linear Formula | (CH3)2CHCH2OH |
Quantity | 500 mL |
UN Number | UN1212 |
Beilstein | 1730878 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'