missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-propanol, 99.5%, Honeywell™
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
CAS | 78-83-1 |
Melting Point | -108°C |
Color | Colorless |
Density | 0.803g/mL at 25°C |
Boiling Point | 108°C |
Flash Point | 28°C (82.4°F) |
Ignition Point | 801°F |
Odor | Characteristic |
Packaging | Glass bottle |
Molecular Formula | C4H10O |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'