missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propanol, 99+%, ACS reagent, Thermo Scientific Chemicals

£27.44 - £82.41

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10311211
View Documents Promotion Details
Thermo Scientific Acros
412655000
500 mL Glass Bottle
£27.44
500mL
Please to purchase this item. Need a web account? Register with us today!
10698994
View Documents
Thermo Scientific Acros
412650025
2.5 L Glass bottle
£82.41
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78-83-1
74.12
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
6560
2-methylpropan-1-ol
C4H10O
MFCD00004740
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
CHEBI:46645
CC(C)CO

Specifications

78-83-1
0.8000g/mL
28°C
99% min. (GC)
(CH3)2CHCH2OH
MFCD00004740
0.001% max.
0.8
(in water) Passes test
CC(C)CO
74.12
CHEBI:46645
74.12
99+%
0.0005 meq/g max
2-Methyl-1-propanol, 99+%
-108°C
108°C
Authentic
C4H10O
1.3945 to 1.3965
01, 373
15, 5176
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
2-methylpropan-1-ol
6560
4 Pas (20°C)
11.7mbar at 20°C
ACS Reagent
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-148-0

missing translation for 'rtecsNumber' : NP9625000

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Methyl-1-propanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS
Documents

Documents

Certificates

RUO – Research Use Only