missing translation for 'onlineSavingsMsg'
Learn More
2-Methyl-1-propanol, 99+%, Extra Pure, Thermo Scientific Chemicals Product Code.: 10317860

2-Methyl-1-propanol, 99+%, Extra Pure, Thermo Scientific Chemicals

Product Code. 10317860
1 L, Glass bottle
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
This item is not returnable. View return policy

Product Code. 10317860

Brand: Thermo Scientific Acros 158280010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Methyl-1-propanol, 99+%, C4H10O, CAS Number-78-83-1, 1-hydroxymethylpropane, 1-propanol, 2-methyl, 2-methyl-1-propanol, 2-methylpropyl alcohol, i-butyl alcohol, isobutanol, iso-butyl alcohol, isobutyl alcohol, isobutylalkohol, isopropylcarbinol, 2.5L, 99% min. (GC), 01, 373, 108 deg.C, 100.0, 99.0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcoholShow MoreShow Less
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -108°C
Density 0.8000g/mL
Boiling Point 108°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2OH
Refractive Index 1.3945 to 1.3965
Quantity 1 L
Beilstein 01, 373
Residue after Evaporation 0.005% max.
Merck Index 15, 5176
Specific Gravity 0.8
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone
Viscosity 4 Pas (20°C)
Formula Weight 74.12
Vapor Pressure 11.7mbar at 20°C
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-propanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468638 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number158280250,158280010,158280025
Lot NumberA0461029 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number158280250,158280010,158280025
Lot NumberA0457776 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number158280250,158280010,158280025
Lot NumberA0402561 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number158280250,158280010,158280025
Lot NumberA0411600 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number158280250,158280010,158280025
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-1-propanol, 99+%, Extra Pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.