Learn More
2-Methyl-1-propanol, 99+%, spectrophotometric grade, Thermo Scientific Chemicals
£48.20 - £184.05
Chemical Identifiers
CAS | 78-83-1 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00004740 |
InChI Key | ZXEKIIBDNHEJCQ-UHFFFAOYSA-N |
Synonym | 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol |
PubChem CID | 6560 |
ChEBI | CHEBI:46645 |
IUPAC Name | 2-methylpropan-1-ol |
SMILES | CC(C)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10514111
|
Thermo Scientific Acros
167705000 |
500 mL | Glass bottle |
£48.20
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10357920
|
Thermo Scientific Acros
167700010 |
1 L | Glass bottle |
£88.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10367920
|
Thermo Scientific Acros
167700025 |
2.5 L | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
78-83-1 | |
74.12 | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
6560 | |
2-methylpropan-1-ol |
C4H10O | |
MFCD00004740 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
CHEBI:46645 | |
CC(C)CO |
Specifications
78-83-1 | |
99.0 | |
-108.0°C | |
108.0°C | |
99% min. (GC) | |
(CH3)2CHCH2OH | |
MFCD00004740 | |
0.0005% max. | |
0.8 | |
Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone | |
CC(C)CO | |
74.12 | |
CHEBI:46645 | |
74.12 | |
Liquid |
0.01 max. at 330nm, 0.1 max. at 250nm, 0.5 max. at 230nm, 1 max. at 220nm | |
100.0 | |
0.8000g/mL | |
28°C | |
C4H10O | |
1.3945 to 1.3965 | |
01, 373 | |
15, 5176 | |
2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
2-methylpropan-1-ol | |
6560 | |
4 Pas (20°C) | |
99+% | |
2-Methyl-1-propanol, 99+% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-148-0
missing translation for 'rtecsNumber' : NP9625000
missing translation for 'tsca' : TSCA
- 2-Methyl-1-propanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only