missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-(4-methylphenyl)morpholine, 97%, Thermo Scientific Chemicals

Product Code. 11494650
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-81-1
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD08060961
InChI Key NQYMBSUCKZVKFF-UHFFFAOYSA-N
Synonym 2-methyl-2-p-tolyl morpholine, 2-methyl-2-4-methylphenyl morpholine, 2-methyl-2-p-tolylmorpholine, 2-methyl-2-p-tolyl-morpholine, 2-4-methylphenyl-2-methylmorpholine, morpholine, 2-methyl-2-4-methylphenyl
PubChem CID 24208807
IUPAC Name 2-methyl-2-(4-methylphenyl)morpholine
SMILES CC1=CC=C(C=C1)C2(CNCCO2)C

Specifications

Melting Point 49°C to 51°C
Quantity 250 mg
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-(4-methylphenyl)morpholine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.