missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butanol, 98%, Thermo Scientific Chemicals

Product Code. 11443857
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.

Solubility
Miscible with alcohol, ether, benzene, chloroform, glycerol, oils and acetone.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-85-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004478
InChI Key MSXVEPNJUHWQHW-UHFFFAOYSA-N
Synonym 2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
PubChem CID 6405
IUPAC Name 2-methylbutan-2-ol
SMILES CCC(C)(C)O

Specifications

Melting Point -12°C
Density 0.805 g/mL
Boiling Point 101°C to 103°C
Flash Point 20°C (68°F)
Assay Percent Range 0.98
Refractive Index 1.405
Quantity 2500 mL
UN Number UN1105
Beilstein 1361351
Merck Index 14,7140
Solubility Information Miscible with alcohol,ether,benzene,chloroform,glycerol,oils and acetone.
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-butanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.