Learn More
2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene, Thermo Scientific Chemicals
£75.80 - £640.00
Chemical Identifiers
CAS | 513-35-9 |
---|---|
Molecular Formula | C5H10 |
Molecular Weight (g/mol) | 70.14 |
MDL Number | MFCD00009276 |
InChI Key | BKOOMYPCSUNDGP-UHFFFAOYSA-N |
Synonym | 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2 |
PubChem CID | 10553 |
ChEBI | CHEBI:77916 |
IUPAC Name | 2-methylbut-2-ene |
SMILES | CC=C(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10196673
|
Thermo Scientific Acros
414092500 |
250 mL | Glass bottle |
£75.80
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10453385
|
Thermo Scientific
414090010 |
1 L | Glass bottle |
£320.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11378621
|
Thermo Scientific Acros
414090025 |
2.5 L | Glass bottle |
£640.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
513-35-9 | |
70.14 | |
BKOOMYPCSUNDGP-UHFFFAOYSA-N | |
10553 | |
2-methylbut-2-ene |
C5H10 | |
MFCD00009276 | |
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2 | |
CHEBI:77916 | |
CC=C(C)C |
Specifications
513-35-9 | |
Colorless to Yellow | |
35.0°C to 38.0°C | |
Authentic | |
C5H10 | |
1.3860 to 1.3880 | |
01,211 | |
0.66 | |
BKOOMYPCSUNDGP-UHFFFAOYSA-N | |
2-methylbut-2-ene | |
10553 | |
70.13 | |
Liquid |
-134.0°C | |
0.6600g/mL | |
−45°C | |
Balance mainly 2-methyl-1-butene (GC) | |
CH3CH=C(CH3)2 | |
MFCD00009276 | |
15,601 | |
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2 | |
CC=C(C)C | |
70.14 | |
CHEBI:77916 | |
90% | |
2-Methyl-2-butene |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected o
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a well-ventilated place.
Keep cool.
Keep away from heat/sparks/open flame
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 208-156-3
RUO – Research Use Only