missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-propen-1-ol, 98%, Thermo Scientific Chemicals

Product Code. 15406978
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-42-8
Molecular Formula C4H8O
MDL Number MFCD00004737
InChI Key BYDRTKVGBRTTIT-UHFFFAOYSA-N
PubChem CID 10557
IUPAC Name 2-methylprop-2-en-1-ol
SMILES CC(=C)CO

Specifications

Density 0.857
Boiling Point 113°C to 115°C
Flash Point 32°C (90°F)
Linear Formula CH2=C(CH3)CH2OH
Refractive Index 1.426
Quantity 5 g
UN Number UN2614
Beilstein 969226
Synonym methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
Molecular Weight (g/mol) 72.107
Formula Weight 72.11
Percent Purity 98%
Chemical Name or Material 2-Methyl-2-propen-1-ol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.