missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2h-indazole-3-carboxylic acid, 97%, Thermo Scientific™

£44.90 - £316.00

Chemical Identifiers

CAS 34252-44-3
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
InChI Key ABRISPFTOZIXMP-UHFFFAOYSA-N
Synonym 2-methyl-2h-indazole-3-carboxylic acid, 2h-indazole-3-carboxylic acid, 2-methyl
PubChem CID 7060537
IUPAC Name 2-methylindazole-3-carboxylic acid
SMILES CN1C(=C2C=CC=CC2=N1)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10601665
View Documents
Thermo Scientific Maybridge
CC44401CB
250 mg
£44.90
250mg
Please to purchase this item. Need a web account? Register with us today!
10309902
View Documents
Thermo Scientific Maybridge
CC44401DA
1 g
£136.00
1g
Please to purchase this item. Need a web account? Register with us today!
10492654
View Documents
Thermo Scientific Maybridge
CC44401DE
5 g
£316.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

34252-44-3
176.175
2-methyl-2h-indazole-3-carboxylic acid, 2h-indazole-3-carboxylic acid, 2-methyl
2-methylindazole-3-carboxylic acid
C9H8N2O2
ABRISPFTOZIXMP-UHFFFAOYSA-N
7060537
CN1C(=C2C=CC=CC2=N1)C(=O)O

Specifications

34252-44-3
C9H8N2O2
ABRISPFTOZIXMP-UHFFFAOYSA-N
2-methylindazole-3-carboxylic acid
7060537
97%
97%
2-methyl-2h-indazole-3-carboxylic acid, 2h-indazole-3-carboxylic acid, 2-methyl
CN1C(=C2C=CC=CC2=N1)C(=O)O
176.175
176.17
2-Methyl-2H-indazole-3-carboxylic acid
Videos
SDS
Documents

Documents

Product Certifications