missing translation for 'onlineSavingsMsg'
Learn More
2-Methyl-3,4,5,6,7,8-hexahydrobenzo[4,5]thieno[2,3-d]pyrimidine-4-thione, 96%, Thermo Scientific™ Product Code.: 15415185

2-Methyl-3,4,5,6,7,8-hexahydrobenzo[4,5]thieno[2,3-d]pyrimidine-4-thione, 96%, Thermo Scientific™

Product Code. 15415185
500 mg
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy

Product Code. 15415185

Brand: Thermo Scientific Alfa Aesar H33975.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 23922-04-5
Melting Point >250°C
Molecular Formula C11H12N2S2
MDL Number MFCD00447466
Quantity 500 mg
Synonym 2-methyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine-4-thiol, 1 benzothieno 2,3-d pyrimidine-4 3h-thione,5,6,7,8-tetrahydro-2-methyl, 2-methyl-3,4,5,6,7,8-hexahydrobenzo 4,5 thieno 2,3-d pyrimidine-4-thione, 5-methyl-8-thia-4,6-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,5-triene-3-thione, 2-methyl-3,5,6,7,8-pentahydrobenzo b thiopheno 2,3-d pyrimidine-4-thione, 2-methyl-5,6,7,8-tetrahydro-3h-1 benzothiolo 2,3-d pyrimidine-4-thione, 2-methyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine-4 3h-thione, 2-methyl-5,6,7,8-tetrahydrobenzo 4,5 thieno 2,3-d pyrimidine-4-thiol, 2-methyl-5,6,7,8-tetrahydrobenzo b thieno 2,3-d pyrimidine-4 3h-thione, 2-methyl-5,6,7,8-tetrahydrobenzo b thiopheno 2,3-d pyrimidine-4-thiol
InChI Key YLQYIQCHQZYLQB-UHFFFAOYSA-N
SMILES CC1=NC2=C(C3=C(S2)CCCC3)C(=S)N1
IUPAC Name 2-methyl-5,6,7,8-tetrahydro-3H-[1]benzothiolo[2,3-d]pyrimidine-4-thione
Molecular Weight (g/mol) 236.351
PubChem CID 699511
Formula Weight 236.36
Percent Purity 96%
Chemical Name or Material 2-Methyl-3,4,5,6,7,8-hexahydrobenzo[4,5]thieno[2,3-d]pyrimidine-4-thione
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-3,4,5,6,7,8-hexahydrobenzo[4,5]thieno[2,3-d]pyrimidine-4-thione, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.