missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-isothiazolin-3-one, 50% aqueous solution, Thermo Scientific Chemicals

£51.66 - £195.98

Chemical Identifiers

CAS 2682-20-4
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
InChI Key BEGLCMHJXHIJLR-UHFFFAOYSA-N
Synonym 2-methyl-4-isothiazolin-3-one, methylisothiazolinone, 2-methyl-4-isothiazoline-3-one, 2-methylisothiazol-3 2h-one, 2-methyl-3 2h-isothiazolone, n-methylisothiazolone, 3 2h-isothiazolone, 2-methyl, caswell no. 572a, 2-methyl-3-isothiazolone, unii-229d0e1qfa
PubChem CID 39800
ChEBI CHEBI:53620
IUPAC Name 2-methyl-1,2-thiazol-3-one
SMILES CN1C(=O)C=CS1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15589503
View Documents Promotion Details
Thermo Scientific Acros
459970050
5 g
£51.66
5g
Please to purchase this item. Need a web account? Register with us today!
15599503
View Documents
Thermo Scientific Acros
459970250
25 g
£195.98
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2682-20-4
115.15
2-methyl-4-isothiazolin-3-one, methylisothiazolinone, 2-methyl-4-isothiazoline-3-one, 2-methylisothiazol-3 2h-one, 2-methyl-3 2h-isothiazolone, n-methylisothiazolone, 3 2h-isothiazolone, 2-methyl, caswell no. 572a, 2-methyl-3-isothiazolone, unii-229d0e1qfa
CHEBI:53620
CN1C(=O)C=CS1
C4H5NOS
BEGLCMHJXHIJLR-UHFFFAOYSA-N
39800
2-methyl-1,2-thiazol-3-one

Specifications

2682-20-4
5.0 to 7.0 (1% aq. soln.)
1.16
BEGLCMHJXHIJLR-UHFFFAOYSA-N
2-methyl-1,2-thiazol-3-one
39800
115.15
1.1600g/mL
C4H5NOS
2-methyl-4-isothiazolin-3-one, methylisothiazolinone, 2-methyl-4-isothiazoline-3-one, 2-methylisothiazol-3 2h-one, 2-methyl-3 2h-isothiazolone, n-methylisothiazolone, 3 2h-isothiazolone, 2-methyl, caswell no. 572a, 2-methyl-3-isothiazolone, unii-229d0e1qfa
CN1C(=O)C=CS1
115.15
CHEBI:53620
2-Methyl-4-isothiazolin-3-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic if inhaled.
May cause respiratory irritation.
Very toxic to aquatic life.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 220-239-6

Product Identifier
  • 2-Methyl-4-isothiazolin-3-one, 50% aqueous solution

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only