missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-n-propyl-1,3-oxathiane, cis + trans, 99%, Thermo Scientific™

Product Code. 11465237
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67715-80-4
Molecular Formula C8H16OS
Molecular Weight (g/mol) 160.275
MDL Number MFCD00673203
InChI Key GKGOLPMYJJXRGD-UHFFFAOYSA-N
Synonym 1,3-oxathiane, 2-methyl-4-propyl, 2-methyl-4-propyl-1,3-oxathiane, mixture of cis and trans, 1,3-oxathiane, 2-methyl-4-propyl-, 2r,4r-rel, 1,3-oxathiane, 2-methyl-4-propyl-, 2r,4s-rel, tropathiane, z-2-methyl-4-propyl-1,3-oxathiane, 2-tropical oxathiane, e-galbanum oxathiane, z-galbanum oxathiane, dsstox_cid_27704
PubChem CID 101010
ChEBI CHEBI:87335
IUPAC Name 2-methyl-4-propyl-1,3-oxathiane
SMILES CCCC1CCOC(S1)C

Specifications

Boiling Point 218°C to 220°C
Quantity 5 g
UN Number UN1993
Formula Weight 160.27
Percent Purity 99%
Chemical Name or Material 2-Methyl-4-n-propyl-1,3-oxathiane, cis + trans
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-4-n-propyl-1,3-oxathiane, cis + trans, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.