missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Methyl-4'-nitroacetanilide, 97%, Thermo Scientific™
£148.00
Chemical Identifiers
CAS | 2719-15-5 |
---|---|
Molecular Formula | C9H10N2O3 |
Molecular Weight (g/mol) | 194.19 |
MDL Number | MFCD00033896 |
InChI Key | JZEOVPGWIWSSAK-UHFFFAOYSA-N |
Synonym | n-2-methyl-4-nitrophenyl acetamide, 4'-nitro-o-acetotoluidide, n1-2-methyl-4-nitrophenyl acetamide, acetamide, n-2-methyl-4-nitrophenyl, 2'-methyl-4'-nitroacetanilide, acetyl-p-nitro-o-toluidine, 2-methyl-4-nitroacetanilide, o-acetotoluidide, 4'-nitro, acetamide,n-2-methyl-4-nitrophenyl, maybridge1_001434 |
PubChem CID | 75936 |
IUPAC Name | N-(2-methyl-4-nitrophenyl)acetamide |
SMILES | CC(=O)NC1=CC=C(C=C1C)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11489547
|
Thermo Scientific Alfa Aesar
B22054.14 |
25 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2′-Methyl-4′-nitroacetanilide acts as the reactant in the synthesis of TNT isomers.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, bases.
Chemical Identifiers
2719-15-5 | |
194.19 | |
JZEOVPGWIWSSAK-UHFFFAOYSA-N | |
75936 | |
CC(=O)NC1=CC=C(C=C1C)[N+]([O-])=O |
C9H10N2O3 | |
MFCD00033896 | |
n-2-methyl-4-nitrophenyl acetamide, 4'-nitro-o-acetotoluidide, n1-2-methyl-4-nitrophenyl acetamide, acetamide, n-2-methyl-4-nitrophenyl, 2'-methyl-4'-nitroacetanilide, acetyl-p-nitro-o-toluidine, 2-methyl-4-nitroacetanilide, o-acetotoluidide, 4'-nitro, acetamide,n-2-methyl-4-nitrophenyl, maybridge1_001434 | |
N-(2-methyl-4-nitrophenyl)acetamide |
Specifications
2719-15-5 | |
C9H10N2O3 | |
n-2-methyl-4-nitrophenyl acetamide, 4'-nitro-o-acetotoluidide, n1-2-methyl-4-nitrophenyl acetamide, acetamide, n-2-methyl-4-nitrophenyl, 2'-methyl-4'-nitroacetanilide, acetyl-p-nitro-o-toluidine, 2-methyl-4-nitroacetanilide, o-acetotoluidide, 4'-nitro, acetamide,n-2-methyl-4-nitrophenyl, maybridge1_001434 | |
CC(=O)NC1=CC=C(C=C1C)[N+]([O-])=O | |
194.19 | |
194.19 | |
2'-Methyl-4'-nitroacetanilide |
198°C to 200°C | |
MFCD00033896 | |
JZEOVPGWIWSSAK-UHFFFAOYSA-N | |
N-(2-methyl-4-nitrophenyl)acetamide | |
75936 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures