Learn More
2-Methyl-5-(methylthio)furan, 99%, Thermo Scientific Chemicals
£44.30 - £148.00
Chemical Identifiers
CAS | 13678-59-6 |
---|---|
Molecular Formula | C6H8OS |
Molecular Weight (g/mol) | 128.189 |
MDL Number | MFCD01208018 |
InChI Key | RESBOJMQOGJOMW-UHFFFAOYSA-N |
Synonym | 2-methyl-5-methylthio furan, 5-methyl-2-methylthio furan, furan, 2-methyl-5-methylthio, methyl 5-methylfuryl sulfide, 2-methyl-5-methylsulfanyl furan, 2-methyl-5-methylthiofuran, unii-41z7b950jj, 2-methyl 5-methyl thio furan, furan,2-methyl-5-methylthio, fema no. 3366 |
PubChem CID | 61657 |
IUPAC Name | 2-methyl-5-methylsulfanylfuran |
SMILES | CC1=CC=C(O1)SC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11459477
|
Thermo Scientific Alfa Aesar
B21998.04 |
2 g |
£44.30
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469477
|
Thermo Scientific Alfa Aesar
B21998.09 |
10 g |
£148.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13678-59-6 | |
128.189 | |
RESBOJMQOGJOMW-UHFFFAOYSA-N | |
61657 | |
CC1=CC=C(O1)SC |
C6H8OS | |
MFCD01208018 | |
2-methyl-5-methylthio furan, 5-methyl-2-methylthio furan, furan, 2-methyl-5-methylthio, methyl 5-methylfuryl sulfide, 2-methyl-5-methylsulfanyl furan, 2-methyl-5-methylthiofuran, unii-41z7b950jj, 2-methyl 5-methyl thio furan, furan,2-methyl-5-methylthio, fema no. 3366 | |
2-methyl-5-methylsulfanylfuran |
Specifications
13678-59-6 | |
79°C to 81°C (50 mmHg) | |
Stench | |
MFCD01208018 | |
2-methyl-5-methylthio furan, 5-methyl-2-methylthio furan, furan, 2-methyl-5-methylthio, methyl 5-methylfuryl sulfide, 2-methyl-5-methylsulfanyl furan, 2-methyl-5-methylthiofuran, unii-41z7b950jj, 2-methyl 5-methyl thio furan, furan,2-methyl-5-methylthio, fema no. 3366 | |
CC1=CC=C(O1)SC | |
128.189 | |
128.19 | |
2-Methyl-5-(methylthio)furan |
1.063 | |
50°C (122°F) | |
C6H8OS | |
UN1993 | |
RESBOJMQOGJOMW-UHFFFAOYSA-N | |
2-methyl-5-methylsulfanylfuran | |
61657 | |
99% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 237-170-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only