missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-5-phenyl-3-furyl isocyanate, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 199.21 |
Percent Purity | 95% |
Chemical Name or Material | 2-Methyl-5-phenyl-3-furyl isocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'