missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethyl)phenol, 97%, Thermo Scientific™

Catalog Number 15454367
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1017778-05-0
C8H7F3O
176.14
MFCD09832287
CBWYZBGMPOIRBC-UHFFFAOYSA-N
2-methyl-5-trifluoromethyl phenol, phenol, 2-methyl-5-trifluoromethyl, 3-hydroxy-4-methylbenzotrifluoride, 3-hydroxy-4-trifluoromethyl toluene
20111828
2-methyl-5-(trifluoromethyl)phenol
CC1=C(O)C=C(C=C1)C(F)(F)F
This item is not returnable. View return policy
1017778-05-0
C8H7F3O
176.14
MFCD09832287
CBWYZBGMPOIRBC-UHFFFAOYSA-N
2-methyl-5-trifluoromethyl phenol, phenol, 2-methyl-5-trifluoromethyl, 3-hydroxy-4-methylbenzotrifluoride, 3-hydroxy-4-trifluoromethyl toluene
20111828
2-methyl-5-(trifluoromethyl)phenol
CC1=C(O)C=C(C=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3265
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-(trifluoromethyl)phenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'