missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-piperazin-1-ylpyrazine, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 2-methyl-6-piperazin-1-yl pyrazine, 2-methyl-6-piperazinylpyrazine, pyrazine,2-methyl-6-1-piperazinyl, pyrazine, 2-methyl-6-1-piperazinyl, pyrazine, 2-methyl-6-1-piperazinyl-9ci |
Formula Weight | 178.24 |
Percent Purity | 95% |
Chemical Name or Material | 2-Methyl-6-piperazin-1-ylpyrazine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'