missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-piperazin-1-ylpyrazine, ≥95%, Thermo Scientific™

Product Code. 10737675
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51047-59-7
Molecular Formula C9H14N4
Molecular Weight (g/mol) 178.239
MDL Number MFCD08272850
InChI Key ZQAVHWZTBHGKFK-UHFFFAOYSA-N
PubChem CID 20290698
IUPAC Name 2-methyl-6-piperazin-1-ylpyrazine
SMILES CC1=CN=CC(=N1)N2CCNCC2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-methyl-6-piperazin-1-yl pyrazine, 2-methyl-6-piperazinylpyrazine, pyrazine,2-methyl-6-1-piperazinyl, pyrazine, 2-methyl-6-1-piperazinyl, pyrazine, 2-methyl-6-1-piperazinyl-9ci
Formula Weight 178.24
Percent Purity 95%
Chemical Name or Material 2-Methyl-6-piperazin-1-ylpyrazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'