missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Thermo Scientific™
£226.45 - £676.24
Chemical Identifiers
CAS | 1257535-05-9 |
---|---|
Molecular Formula | C7H5ClF3NO2S |
Molecular Weight (g/mol) | 259.63 |
MDL Number | MFCD14583150 |
InChI Key | BNCNLCKWRCXBQH-UHFFFAOYSA-N |
Synonym | 2-methyl-6-trifluoromethyl pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-2-methyl-6-trifluoromethyl pyridine |
PubChem CID | 51342031 |
IUPAC Name | 2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride |
SMILES | CC1=C(C=CC(=N1)C(F)(F)F)S(Cl)(=O)=O |
Description
Chemical Identifiers
1257535-05-9 | |
259.63 | |
BNCNLCKWRCXBQH-UHFFFAOYSA-N | |
51342031 | |
CC1=C(C=CC(=N1)C(F)(F)F)S(Cl)(=O)=O |
C7H5ClF3NO2S | |
MFCD14583150 | |
2-methyl-6-trifluoromethyl pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-2-methyl-6-trifluoromethyl pyridine | |
2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride |
Specifications
1257535-05-9 | |
97% | |
C7H5ClF3NO2S | |
2-methyl-6-trifluoromethyl pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-2-methyl-6-trifluoromethyl pyridine | |
CC1=C(C=CC(=N1)C(F)(F)F)S(Cl)(=O)=O | |
259.63 | |
259.64 | |
2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride |
Authentic | |
Glass Bottle | |
MFCD14583150 | |
BNCNLCKWRCXBQH-UHFFFAOYSA-N | |
2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride | |
51342031 | |
97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
Wear face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin wi
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only