missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)butane hydrochloride, 94%, Thermo Scientific Chemicals

Product Code. 11441840
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hydrochloric acid.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75098-40-7
Molecular Formula C5H14ClN
Molecular Weight (g/mol) 123.624
MDL Number MFCD07777098
InChI Key ICBNZSHUWSBMCL-UHFFFAOYSA-N
Synonym n-methyl-sec-butylamine hydrochloride, 2-methylamino butane hydrochloride, methyl sec-butyl amine hydrochloride, butan-2-yl methyl amine hydrochloride, n-methyl-2-butanamine hydrochloride, n-methylbutan-2-amine hydrochloride, n-methyl-2-butylamine hydrochloride, n-methylbutan-2-amine-hydrogen chloride 1/1
PubChem CID 21273362
IUPAC Name N-methylbutan-2-amine;hydrochloride
SMILES CCC(C)NC.Cl

Specifications

Melting Point 76°C to 82°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in hydrochloric acid.
Formula Weight 123.62
Percent Purity 94%
Chemical Name or Material 2-(Methylamino)butane hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'