missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Methylamino)butane hydrochloride, 94%, Thermo Scientific Chemicals
£46.20 - £146.00
Chemical Identifiers
CAS | 75098-40-7 |
---|---|
Molecular Formula | C5H14ClN |
Molecular Weight (g/mol) | 123.624 |
MDL Number | MFCD07777098 |
InChI Key | ICBNZSHUWSBMCL-UHFFFAOYSA-N |
Synonym | n-methyl-sec-butylamine hydrochloride, 2-methylamino butane hydrochloride, methyl sec-butyl amine hydrochloride, butan-2-yl methyl amine hydrochloride, n-methyl-2-butanamine hydrochloride, n-methylbutan-2-amine hydrochloride, n-methyl-2-butylamine hydrochloride, n-methylbutan-2-amine-hydrogen chloride 1/1 |
PubChem CID | 21273362 |
IUPAC Name | N-methylbutan-2-amine;hydrochloride |
SMILES | CCC(C)NC.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11441840
|
Thermo Scientific Alfa Aesar
H27547.03 |
1 g |
£46.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11451840
|
Thermo Scientific Alfa Aesar
H27547.06 |
5 g |
£146.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in hydrochloric acid.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
75098-40-7 | |
123.624 | |
ICBNZSHUWSBMCL-UHFFFAOYSA-N | |
21273362 | |
CCC(C)NC.Cl |
C5H14ClN | |
MFCD07777098 | |
n-methyl-sec-butylamine hydrochloride, 2-methylamino butane hydrochloride, methyl sec-butyl amine hydrochloride, butan-2-yl methyl amine hydrochloride, n-methyl-2-butanamine hydrochloride, n-methylbutan-2-amine hydrochloride, n-methyl-2-butylamine hydrochloride, n-methylbutan-2-amine-hydrogen chloride 1/1 | |
N-methylbutan-2-amine;hydrochloride |
Specifications
75098-40-7 | |
C5H14ClN | |
Hygroscopic | |
Soluble in hydrochloric acid. | |
CCC(C)NC.Cl | |
123.624 | |
123.62 | |
2-(Methylamino)butane hydrochloride |
76°C to 82°C | |
MFCD07777098 | |
n-methyl-sec-butylamine hydrochloride, 2-methylamino butane hydrochloride, methyl sec-butyl amine hydrochloride, butan-2-yl methyl amine hydrochloride, n-methyl-2-butanamine hydrochloride, n-methylbutan-2-amine hydrochloride, n-methyl-2-butylamine hydrochloride, n-methylbutan-2-amine-hydrogen chloride 1/1 | |
ICBNZSHUWSBMCL-UHFFFAOYSA-N | |
N-methylbutan-2-amine;hydrochloride | |
21273362 | |
94% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only