Learn More
2-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
£19.00 - £80.80
Chemical Identifiers
CAS | 16419-60-6 |
---|---|
Molecular Formula | C7H9BO2 |
Molecular Weight (g/mol) | 135.96 |
MDL Number | MFCD00093526 |
InChI Key | NSJVYHOPHZMZPN-UHFFFAOYSA-N |
Synonym | 2-tolylboronic acid, o-tolylboronic acid, 2-methylbenzeneboronic acid, 2-methylphenyl boronic acid, o-methylphenylboronic acid, 2-methylphenyl boranediol, tolylboronic acid, o-tolueneboronic acid, toluene-2-boronic acid, boronic acid, methylphenyl |
PubChem CID | 2733267 |
IUPAC Name | (2-methylphenyl)boronic acid |
SMILES | CC1=CC=CC=C1B(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11436386
|
Thermo Scientific Alfa Aesar
B23154.03 |
1 g |
£19.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11446386
|
Thermo Scientific Alfa Aesar
B23154.06 |
5 g |
£31.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11456386
|
Thermo Scientific Alfa Aesar
B23154.14 |
25 g |
£80.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
Chemical Identifiers
16419-60-6 | |
135.96 | |
NSJVYHOPHZMZPN-UHFFFAOYSA-N | |
2733267 | |
CC1=CC=CC=C1B(O)O |
C7H9BO2 | |
MFCD00093526 | |
2-tolylboronic acid, o-tolylboronic acid, 2-methylbenzeneboronic acid, 2-methylphenyl boronic acid, o-methylphenylboronic acid, 2-methylphenyl boranediol, tolylboronic acid, o-tolueneboronic acid, toluene-2-boronic acid, boronic acid, methylphenyl | |
(2-methylphenyl)boronic acid |
Specifications
16419-60-6 | |
C7H9BO2 | |
2935796 | |
Slightly soluble in water. | |
CC1=CC=CC=C1B(O)O | |
135.96 | |
135.96 | |
2-Methylbenzeneboronic acid |
∼162°C to 164°C | |
MFCD00093526 | |
2-tolylboronic acid, o-tolylboronic acid, 2-methylbenzeneboronic acid, 2-methylphenyl boronic acid, o-methylphenylboronic acid, 2-methylphenyl boranediol, tolylboronic acid, o-tolueneboronic acid, toluene-2-boronic acid, boronic acid, methylphenyl | |
NSJVYHOPHZMZPN-UHFFFAOYSA-N | |
(2-methylphenyl)boronic acid | |
2733267 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'rtecsNumber' : ED7777777
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only