missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzothiazol-6-ol, 96%, Thermo Scientific Chemicals

Product Code. 15496677
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68867-18-5
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00227234
InChI Key ROFBPPIQUBJMRO-UHFFFAOYSA-N
Synonym 2-methyl-6-benzothiazolol, 2-methylbenzo d thiazol-6-ol, 6-benzothiazolol, 2-methyl, 2-methylbenzothiazol-6-ol, 6-hydroxy-2-methylbenzothiazole, 6-hydroxy-2-methylbenzthiazole, pubchem17329, 2-methyl-benzothiazol-6-ol, 2-methyl-6-hydroxybenzothiazole, 6-hydroxy-2-methyl-benzothiazole
PubChem CID 759304
IUPAC Name 2-methyl-1,3-benzothiazol-6-ol
SMILES CC1=NC2=CC=C(O)C=C2S1

Specifications

Melting Point 159°C to 161°C
Quantity 250 mg
Formula Weight 165.21
Percent Purity 96%
Chemical Name or Material 2-Methylbenzothiazol-6-ol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'