missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzothiazole, 99%, Thermo Scientific Chemicals

Product Code. 15204644
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-75-2
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
InChI Key DXYYSGDWQCSKKO-UHFFFAOYSA-N
Synonym 2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w
PubChem CID 8446
IUPAC Name 2-methyl-1,3-benzothiazole
SMILES CC1=NC2=CC=CC=C2S1

Specifications

CAS Min % 98.5
CAS Max % 99.0
Melting Point 12.0°C to 14.0°C
Color Amber to Colorless
Density 1.2000g/mL
Boiling Point 238.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6150 to 1.6180
Quantity 500 g
Beilstein 27,46
Specific Gravity 1.2
Solubility Information Solubility in water: insoluble. Other solubilities: ssluble in ether and chloroform
Formula Weight 149.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylbenzothiazole, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'