Learn More
2-Methylbenzothiazole, 99%, Thermo Scientific Chemicals
£54.50 - £670.00
Chemical Identifiers
CAS | 120-75-2 |
---|---|
Molecular Formula | C8H7NS |
Molecular Weight (g/mol) | 149.21 |
InChI Key | DXYYSGDWQCSKKO-UHFFFAOYSA-N |
Synonym | 2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w |
PubChem CID | 8446 |
IUPAC Name | 2-methyl-1,3-benzothiazole |
SMILES | CC1=NC2=CC=CC=C2S1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15294634
|
Thermo Scientific Acros
150380250 |
25 g | Glass bottle |
£54.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10031840
|
Thermo Scientific Acros
150381000 |
100 g | Glass bottle |
£132.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15204644
|
Thermo Scientific Acros
150385000 |
500 g | Glass bottle |
£670.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
120-75-2 | |
149.21 | |
2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w | |
2-methyl-1,3-benzothiazole |
C8H7NS | |
DXYYSGDWQCSKKO-UHFFFAOYSA-N | |
8446 | |
CC1=NC2=CC=CC=C2S1 |
Specifications
120-75-2 | |
99.0 | |
Amber to Colorless | |
238.0°C | |
Authentic | |
C8H7NS | |
27,46 | |
2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w | |
DXYYSGDWQCSKKO-UHFFFAOYSA-N | |
2-methyl-1,3-benzothiazole | |
8446 | |
99% | |
2-Methylbenzothiazole, 99% |
98.5 | |
12.0°C to 14.0°C | |
1.2000g/mL | |
102°C | |
98.5% min. (GC) | |
1.6150 to 1.6180 | |
1.2 | |
Solubility in water: insoluble. Other solubilities: ssluble in ether and chloroform | |
CC1=NC2=CC=CC=C2S1 | |
149.21 | |
149.21 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-423-3
missing translation for 'rtecsNumber' : DL5600000
missing translation for 'tsca' : TSCA
RUO – Research Use Only