missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzyl chloride, 98+%, Thermo Scientific Chemicals

Product Code. 11405513
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-45-4
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000899
InChI Key VQRBXYBBGHOGFT-UHFFFAOYSA-N
Synonym 2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw
PubChem CID 11093
IUPAC Name 1-(chloromethyl)-2-methylbenzene
SMILES CC1=CC=CC=C1CCl

Specifications

Melting Point 2°C
Density 1.081
Boiling Point 197°C to 199°C
Flash Point 73°C (163°F)
Odor Pungent
Refractive Index 1.541
Quantity 100 g
UN Number UN3265
Beilstein 386016
Merck Index 14,10091
Formula Weight 140.61
Percent Purity ≥98%
Chemical Name or Material 2-Methylbenzyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'